Assay Details
Target Gene Details
Entrez Gene ID: | 6390 |
Gene Name: | succinate dehydrogenase complex iron sulfur subunit B |
Gene Aliases: |
CWS2, IP, PGL4, SDH, SDH1, SDH2, SDHIP |
Location: |
Chr.1:17018722-17054170 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SDHB | NM_003000.2 | NP_002991.2 | ||
AK312056.1 | ||||
BC007840.2 | AAH07840.1 | |||
BF796275.1 | ||||
D10245.1 | BAA01089.1 | |||
DQ403007.1 | ||||
M32246.1 | AAA35708.1 | |||
U17248.1 | AAA81167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2662800 | Chr.1:17034506 - 17036842 on Build GRCh38 | Deletion | SDHB |
nsv428421 | Chr.1:16437838 - 17157486 on Build GRCh38 | Gain+Loss | CROCCP3 LOC105376796 MST1L SDHB LOC105376805 SPATA21 ATP13A2 RNU1-1 RNU1-2 CROCC MIR3675 MFAP2 LOC102723383 FAM231A RNU1-3 ESPNP RNU1-4 PADI2 LOC100132147 CROCCP2 FAM231B NBPF1 MST1P2 NECAP2 |
esv3585358 | Chr.1:17034521 - 17036879 on Build GRCh38 | Loss | SDHB |
nsv834280 | Chr.1:16888218 - 17123006 on Build GRCh38 | Loss | SDHB LOC105376805 ATP13A2 PADI2 RNU1-2 CROCC MFAP2 |
nsv834269 | Chr.1:16855283 - 17038531 on Build GRCh38 | Loss | SDHB LOC105376805 ATP13A2 RNU1-2 CROCC MIR3675 MFAP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |