Assay Details
Target Gene Details
Entrez Gene ID: | 554203 |
Gene Name: | JPX transcript, XIST activator (non-protein coding) |
Gene Aliases: |
DCBALD06, ENOX, LINC00183, NCRNA00183 |
Location: |
Chr.X:73944324-74070384 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JPX | NR_024582.1 | |||
AK056172.1 | ||||
BP871721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2673779 | Chr.X:73974298 - 73976814 on Build GRCh38 | Deletion | JPX |