Assay Details
Target Gene Details
Entrez Gene ID: | 10251 |
Gene Name: | sprouty RTK signaling antagonist 3 |
Gene Aliases: |
spry-3 |
Location: |
Chr.X:155612565-155782459 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRY3 | NM_001304990.1 | NP_001291919.1 | ||
DA102958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | SNORA56 OPN1MW3 F8A2 RAB39B GDI1 TMLHE-AS1 MIR1184-3 F8 IRAK1 UBL4A H2AFB1 CLIC2 MTCP1 TAZ CTAG2 BRCC3 FAM50A LAGE3 EMD OPN1MW2 FAM3A VBP1 SNORA36A H2AFB2 LOC105373386 MIR1184-1 TMLHE G6PD FUNDC2 HCFC1-AS1 FLNA CTAG1A DKC1 MPP1 OPN1MW SNORA70 F8A1 MECP2 CMC4 PLXNA3 LOC107987332 F8A3 MIR1184-2 RPL10 ATP6AP1 FAM223B SLC10A3 TEX28 CH17-340M24.3 SMIM9 DNASE1L1 LOC101927830 SPRY3 TMEM187 TKTL1 MIR6858 OPN1LW CTAG1B MIR3202-1 IKBKG FAM223A HCFC1 MIR718 GAB3 H2AFB3 MIR3202-2 MIR664B |
nsv519485 | Chr.X:155507715 - 155699751 on Build GRCh38 | Loss | SPRY3 TMLHE |
nsv520699 | Chr.X:155545002 - 155699751 on Build GRCh38 | Gain | SPRY3 TMLHE |
nsv471607 | Chr.X:155652457 - 155812331 on Build GRCh38 | Gain+Loss | SPRY3 |
esv2657377 | Chr.X:155641695 - 155958434 on Build GRCh38 | Deletion | SPRY3 |
esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain | BRCC3 LOC101927830 SPRY3 F8A1 CMC4 F8A2 F8A3 MIR1184-2 RAB39B TMLHE-AS1 VBP1 H2AFB2 MIR1184-3 F8 MIR1184-1 TMLHE FUNDC2 SMIM9 H2AFB1 H2AFB3 CLIC2 MTCP1 MPP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |