Assay Details
Target Gene Details
Entrez Gene ID: | 3897 |
Gene Name: | L1 cell adhesion molecule |
Gene Aliases: |
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAM-L1, N-CAML1, NCAM-L1, S10, SPG1 |
Location: |
Chr.X:153861514-153886174 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L1CAM | NM_001278116.1 | NP_001265045.1 | ||
DA122729.1 |
Target Gene Details
Entrez Gene ID: | 100996465 |
Gene Name: | lung carcinoma-associated 10 |
Gene Aliases: |
LCAP |
Location: |
Chr.X:153880672-153888990 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LCA10 | NR_130768.1 | |||
AF514420.1 | AAQ08117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | MIR3202-1 TKTL1 ABCD1 MAGEA1 LOC107983954 LAGE3 L1CAM PNCK AVPR2 FAM58A PDZD4 CTAG1A EMD NAA10 SLC6A8 DUSP9 ZNF275 PLXNB3 BGN RPL10 FAM223A SNORA70 CH17-340M24.3 PLXNA3 TAZ OPN1MW2 GDI1 IKBKG MIR718 DNASE1L1 UBL4A RENBP OPN1MW3 ATP6AP1 BCAP31 TEX28 OPN1LW IDH3G G6PD FAM3A HAUS7 LOC107987332 SLC10A3 ZFP92 LOC105373381 FAM50A LOC105373383 TREX2 OPN1MW LOC105373378 MIR6858 IRAK1 LCA10 SSR4 MECP2 HCFC1 LOC105373377 SRPK3 HCFC1-AS1 TMEM187 LOC105373386 PNMA6A FLNA ATP2B3 MIR3202-2 ARHGAP4 |
nsv469867 | Chr.X:153813281 - 154018926 on Build GRCh38 | Loss | NAA10 RENBP MIR3202-1 IRAK1 LCA10 L1CAM HCFC1 AVPR2 HCFC1-AS1 TMEM187 PDZD4 MIR3202-2 ARHGAP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |