Assay Details
Target Gene Details
Entrez Gene ID: | 4694 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A1 |
Gene Aliases: |
CI-MWFE, MWFE, ZNF183 |
Location: |
Chr.X:119871771-119876666 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA1 | NM_004541.3 | NP_004532.1 | ||
AB451304.1 | ||||
AB451435.1 | ||||
BC000266.1 | AAH00266.1 | |||
BQ680674.1 | ||||
CR456889.1 | CAG33170.1 | |||
U54993.1 | AAD00084.1 | |||
X81900.1 | CAA57489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv7057 | Chr.X:119862293 - 119894925 on Build GRCh38 | Insertion | NDUFA1 RNF113A |
esv2758888 | Chr.X:119823476 - 120195131 on Build GRCh38 | Gain | NDUFA1 RHOXF2 NKAP RNF113A AKAP14 LINC01402 RHOXF2B RHOXF1-AS1 UPF3B RHOXF1 RHOXF1P1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |