Assay Details
Target Gene Details
Entrez Gene ID: | 1184 |
Gene Name: | chloride voltage-gated channel 5 |
Gene Aliases: |
CLC5, CLCK2, ClC-5, DENTS, NPHL1, NPHL2, XLRH, XRN, hCIC-K2 |
Location: |
Chr.X:49922615-50099235 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLCN5 | NM_001127898.3 | NP_001121370.1 | ||
NM_001127899.3 | NP_001121371.1 | |||
NM_001272102.1 | NP_001259031.1 | |||
XM_017029257.1 | XP_016884746.1 | |||
XM_017029258.1 | XP_016884747.1 | |||
AK056560.1 | ||||
AK092049.1 | ||||
BF513571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33289 | Chr.X:49918651 - 49996249 on Build GRCh38 | Loss | CLCN5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |