Assay Details
Target Gene Details
Entrez Gene ID: | 10159 |
Gene Name: | ATPase H+ transporting accessory protein 2 |
Gene Aliases: |
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS |
Location: |
Chr.X:40580964-40606637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6AP2 | NM_005765.2 | NP_005756.2 | ||
AF109363.1 | AAQ13511.1 | |||
AF248966.1 | AAG44564.1 | |||
AF291814.1 | AAM47531.1 | |||
AK075382.1 | BAC11582.1 | |||
AK293510.1 | ||||
AK296614.1 | ||||
AK297007.1 | ||||
AK297760.1 | ||||
AK314633.1 | ||||
AK315948.1 | ||||
AY038990.1 | AAK83467.1 | |||
BC084541.1 | AAH84541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470350 | Chr.X:40540044 - 40624703 on Build GRCh38 | Gain | MPC1L ATP6AP2 |
nsv518965 | Chr.X:40569250 - 40754042 on Build GRCh38 | Gain | CXorf38 MPC1L MED14OS ATP6AP2 MED14 |
esv33829 | Chr.X:40117374 - 40692316 on Build GRCh38 | Loss | CXorf38 MPC1L BCOR LOC101927476 ATP6AP2 MED14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |