Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | SNORA109 MIR6857 MIR98 MIR6895 MIR4536-2 FAM156A MAGED2 PAGE2 ITIH6 FAM104B GNL3L PAGE5 PAGE3 SNORA11 APEX2 GPR173 PHF8 XAGE3 RIBC1 HSD17B10 TRO SMC1A TSR2 IQSEC2 FAM120C LOC100421746 TSPYL2 KDM5C ALAS2 PAGE2B HUWE1 KANTR PFKFB1 MIR6894 FAM156B MIRLET7F2 MTRNR2L10 FOXR2 USP51 WNK3 MAGEH1 MIR4536-1 FGD1 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | SNORA109 MIR6857 MIR98 MIR6895 MIR4536-2 FAM156A MAGED2 PAGE2 ITIH6 FAM104B GNL3L PAGE5 PAGE3 SNORA11 APEX2 GPR173 PHF8 XAGE3 RIBC1 HSD17B10 TRO SMC1A TSR2 IQSEC2 FAM120C LOC100421746 TSPYL2 KDM5C ALAS2 PAGE2B HUWE1 KANTR PFKFB1 MIR6894 FAM156B MIRLET7F2 MTRNR2L10 FOXR2 USP51 WNK3 MAGEH1 MIR4536-1 FGD1 |