Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | MIR6894 MIR6895 PHF8 IQSEC2 HSD17B10 PAGE5 SNORA109 MIR4536-2 TSR2 FAM156A FAM104B MAGED2 MIR6857 TRO MAGEH1 SNORA11 KANTR GPR173 ITIH6 SMC1A PFKFB1 USP51 MIR4536-1 MIR98 FOXR2 MIRLET7F2 KDM5C MTRNR2L10 FGD1 WNK3 GNL3L ALAS2 TSPYL2 LOC100421746 FAM120C PAGE2 APEX2 RIBC1 PAGE3 HUWE1 XAGE3 PAGE2B FAM156B |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | MIR6894 MIR6895 PHF8 IQSEC2 HSD17B10 PAGE5 SNORA109 MIR4536-2 TSR2 FAM156A FAM104B MAGED2 MIR6857 TRO MAGEH1 SNORA11 KANTR GPR173 ITIH6 SMC1A PFKFB1 USP51 MIR4536-1 MIR98 FOXR2 MIRLET7F2 KDM5C MTRNR2L10 FGD1 WNK3 GNL3L ALAS2 TSPYL2 LOC100421746 FAM120C PAGE2 APEX2 RIBC1 PAGE3 HUWE1 XAGE3 PAGE2B FAM156B |