Assay Details
Target Gene Details
Entrez Gene ID: | 10134 |
Gene Name: | B-cell receptor-associated protein 31 |
Gene Aliases: |
6C6-AG, BAP31, CDM, DDCH, DXS1357E |
Location: |
Chr.X:153700492-153724746 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAP31 | NM_001139441.1 | NP_001132913.1 | ||
NM_001139457.2 | NP_001132929.1 | |||
NM_001256447.1 | NP_001243376.1 | |||
NM_005745.7 | NP_005736.3 | |||
AK057613.1 | ||||
AK125631.1 | ||||
AK222494.1 | BAD96214.1 | |||
AK223059.1 | BAD96779.1 | |||
AU132745.1 | ||||
BC014323.1 | AAH14323.1 | |||
BC065292.1 | AAH65292.1 | |||
BE729820.1 | ||||
BX474419.1 | ||||
CX164660.1 | ||||
X81109.1 | CAA57015.1 | |||
Z31696.1 | CAA83501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | MECP2 L1CAM CH17-340M24.3 FAM58A NAA10 FAM223A ZFP92 MAGEA1 ARHGAP4 LOC105373383 UBL4A GDI1 FLNA TKTL1 IDH3G LOC107987332 LOC105373378 TAZ RPL10 BCAP31 ABCD1 LCA10 EMD TEX28 OPN1MW3 FAM3A CTAG1A G6PD DNASE1L1 SSR4 LAGE3 HCFC1-AS1 ATP6AP1 BGN MIR718 SLC10A3 LOC107983954 PLXNA3 LOC105373386 MIR6858 SLC6A8 IRAK1 OPN1LW PNCK MIR3202-2 FAM50A OPN1MW OPN1MW2 ZNF275 LOC105373381 TREX2 DUSP9 TMEM187 SRPK3 HCFC1 PNMA6A ATP2B3 HAUS7 RENBP MIR3202-1 SNORA70 IKBKG LOC105373377 AVPR2 PLXNB3 PDZD4 |
esv21656 | Chr.X:153687433 - 153750282 on Build GRCh38 | Gain+Loss | SLC6A8 BCAP31 ABCD1 PNCK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |