Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | MAGED2 RIBC1 WNK3 MIR4536-2 PAGE2 KDM5C MIR4536-1 MTRNR2L10 PFKFB1 FAM156B MIR6895 PAGE2B SMC1A LOC100421746 MIR6894 ITIH6 MAGEH1 FAM156A HUWE1 ALAS2 MIR98 TSR2 MIR6857 PAGE5 GNL3L APEX2 XAGE3 FOXR2 FAM120C SNORA11 PAGE3 IQSEC2 HSD17B10 KANTR USP51 FAM104B TSPYL2 FGD1 SNORA109 MIRLET7F2 GPR173 TRO PHF8 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | MAGED2 RIBC1 WNK3 MIR4536-2 PAGE2 KDM5C MIR4536-1 MTRNR2L10 PFKFB1 FAM156B MIR6895 PAGE2B SMC1A LOC100421746 MIR6894 ITIH6 MAGEH1 FAM156A HUWE1 ALAS2 MIR98 TSR2 MIR6857 PAGE5 GNL3L APEX2 XAGE3 FOXR2 FAM120C SNORA11 PAGE3 IQSEC2 HSD17B10 KANTR USP51 FAM104B TSPYL2 FGD1 SNORA109 MIRLET7F2 GPR173 TRO PHF8 |