Assay Details
Target Gene Details
Entrez Gene ID: | 3897 |
Gene Name: | L1 cell adhesion molecule |
Gene Aliases: |
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAM-L1, N-CAML1, NCAM-L1, S10, SPG1 |
Location: |
Chr.X:153861514-153886174 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L1CAM | NM_001278116.1 | NP_001265045.1 | ||
DA122729.1 |
Target Gene Details
Entrez Gene ID: | 100996465 |
Gene Name: | lung carcinoma-associated 10 |
Gene Aliases: |
LCAP |
Location: |
Chr.X:153880672-153888990 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LCA10 | NR_130768.1 | |||
AF514420.1 | AAQ08117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | OPN1MW3 CH17-340M24.3 DNASE1L1 MIR3202-2 FAM50A TMEM187 FAM58A LOC105373378 FLNA MIR718 L1CAM LOC105373383 ZFP92 ABCD1 LOC107987332 LOC107983954 HCFC1-AS1 PLXNB3 PDZD4 ATP6AP1 BGN TREX2 LOC105373386 PLXNA3 AVPR2 SLC10A3 IRAK1 OPN1LW OPN1MW TEX28 MIR6858 BCAP31 SSR4 OPN1MW2 ARHGAP4 MIR3202-1 ATP2B3 UBL4A ZNF275 LAGE3 MAGEA1 HAUS7 PNMA6A SNORA70 RENBP TAZ IDH3G DUSP9 GDI1 PNCK G6PD LOC105373377 NAA10 TKTL1 EMD IKBKG CTAG1A MECP2 SRPK3 FAM223A SLC6A8 LOC105373381 LCA10 RPL10 FAM3A HCFC1 |
esv1000302 | Chr.X:153881712 - 153883282 on Build GRCh38 | Gain | LCA10 L1CAM |
nsv469867 | Chr.X:153813281 - 154018926 on Build GRCh38 | Loss | MIR3202-2 TMEM187 AVPR2 IRAK1 L1CAM RENBP HCFC1-AS1 LCA10 PDZD4 NAA10 ARHGAP4 MIR3202-1 HCFC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |