Assay Details
Target Gene Details
Entrez Gene ID: | 28952 |
Gene Name: | coiled-coil domain containing 22 |
Gene Aliases: |
CXorf37, JM1, RTSC2 |
Location: |
Chr.X:49235467-49250526 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC22 | NM_014008.4 | NP_054727.1 | ||
XM_005272599.3 | XP_005272656.1 | |||
AJ005890.1 | CAA06747.1 | |||
AK291976.1 | ||||
AK296911.1 | ||||
AK298264.1 | ||||
AK316375.1 | ||||
BC000972.2 | AAH00972.1 | |||
BC011675.2 | AAH11675.1 | |||
BG912845.1 | ||||
DC340647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | PPP1R3F GAGE10 CCDC22 FOXP3 CACNA1F |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain | GRIPAP1 PLP2 SLC38A5 PIM2 KCND1 TFE3 TBC1D25 PRICKLE3 EBP TIMM17B ERAS FOXP3 RBM3 SYP-AS1 GPKOW PPP1R3F CCDC120 WDR45 HDAC6 FTSJ1 PCSK1N PQBP1 PORCN SYP GATA1 GLOD5 PRAF2 MAGIX SLC35A2 WDR13 CCDC22 WAS SUV39H1 LOC101927635 OTUD5 CACNA1F |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | GRIPAP1 PLP2 SLC38A5 PIM2 SPACA5B SSX1 KCND1 TFE3 TBC1D25 ZNF630-AS1 PRICKLE3 SSX6 EBP TIMM17B SSX3 ZNF182 ERAS FOXP3 SSX5 RBM3 SYP-AS1 GPKOW PPP1R3F CCDC120 SSX9 WDR45 HDAC6 FTSJ1 ZNF630 PCSK1N PQBP1 SPACA5 SSX4B PORCN SYP GATA1 GLOD5 PRAF2 SSX4 MAGIX SLC35A2 WDR13 CCDC22 WAS SUV39H1 LOC101927635 OTUD5 CACNA1F |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | PPP1R3F GAGE10 CCDC22 FOXP3 CACNA1F |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion | CCDC22 FOXP3 CACNA1F |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |