Assay Details
Target Gene Details
Entrez Gene ID: | 29781 |
Gene Name: | non-SMC condensin II complex subunit H2 |
Gene Aliases: |
CAPH2 |
Location: |
Chr.22:50508216-50524780 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCAPH2 | NM_001185011.1 | NP_001171940.1 | ||
NM_014551.4 | NP_055366.3 | |||
NM_152299.3 | NP_689512.2 | |||
XM_005261912.4 | XP_005261969.1 | |||
XM_011530685.2 | XP_011528987.1 | |||
XM_017028793.1 | XP_016884282.1 | |||
AL021682.1 | CAA16670.1 | |||
BC000473.2 | AAH00473.1 | |||
BC001509.2 | AAH01509.1 | |||
BC001833.2 | AAH01833.1 | |||
BC001937.1 | AAH01937.1 | |||
BC009441.1 | AAH09441.1 | |||
CR456604.1 | CAG30490.1 | |||
DB498781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758852 | Chr.22:50491090 - 50578979 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160782 | Chr.22:50463961 - 50571524 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834238 | Chr.22:50402690 - 50556965 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471224 | Chr.22:50435189 - 50580482 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829340 | Chr.22:50505720 - 50603696 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv955201 | Chr.22:50421672 - 50537371 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834239 | Chr.22:50485317 - 50630215 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv589222 | Chr.22:50491152 - 50589773 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs77475245] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)