Assay Details
Target Gene Details
Entrez Gene ID: | 101929372 |
Gene Name: | uncharacterized LOC101929372 |
Gene Aliases: |
- |
Location: |
Chr.22:17580157-17589192 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929372 | NM_001288707.1 | NP_001275636.1 |
Target Gene Details
Entrez Gene ID: | 83733 |
Gene Name: | solute carrier family 25 member 18 |
Gene Aliases: |
GC2 |
Location: |
Chr.22:17562470-17590890 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A18 | NM_001303484.1 | NP_001290413.1 | ||
NM_031481.2 | NP_113669.1 | |||
XM_011546149.2 | XP_011544451.1 | |||
XM_011546150.2 | XP_011544452.1 | |||
XM_011546151.2 | XP_011544453.1 | |||
XM_011546152.1 | XP_011544454.1 | |||
XM_011546153.2 | XP_011544455.1 | |||
XM_011546154.2 | XP_011544456.1 | |||
XM_017028968.1 | XP_016884457.1 | |||
XM_017028969.1 | XP_016884458.1 | |||
XM_017028970.1 | XP_016884459.1 | |||
XM_017028971.1 | XP_016884460.1 | |||
XM_017028972.1 | XP_016884461.1 | |||
XM_017028973.1 | XP_016884462.1 | |||
XM_017028974.1 | XP_016884463.1 | |||
XM_017028975.1 | XP_016884464.1 | |||
AJ428203.1 | CAD21008.1 | |||
AK309110.1 | ||||
AY008285.1 | AAG22855.1 | |||
BC031644.1 | AAH31644.1 | |||
CR456578.1 | CAG30464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4464n100 | Chr.22:17570790 - 17611340 on Build GRCh38 | Gain | ATP6V1E1 LOC101929372 SLC25A18 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | BCL2L13 BID LINC00528 MICAL3 FLJ41941 MIR3198-1 ATP6V1E1 LOC101929372 MIR648 SLC25A18 |
esv3568229 | Chr.22:17574659 - 17596669 on Build GRCh38 | Loss | ATP6V1E1 LOC101929372 SLC25A18 |
nsv3553 | Chr.22:17553601 - 17608096 on Build GRCh38 | Deletion | CECR2 ATP6V1E1 LOC101929372 SLC25A18 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | PI4KAP1 FLJ41941 TMEM191B CECR2 LINC00528 TUBA8 MIR3198-1 ATP6V1E1 LOC101929372 FAM230A BID PEX26 RIMBP3 GGT3P LOC100996432 LOC100996415 MIR648 BCL2L13 LOC642643 MICAL3 LOC729461 LOC100996401 LOC102725072 SLC25A18 USP18 |
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | CECR2 BCL2L13 ATP6V1E1 LOC101929372 SLC25A18 |
nsv1065055 | Chr.22:17559085 - 17632626 on Build GRCh38 | Gain | BCL2L13 ATP6V1E1 LOC101929372 SLC25A18 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | CECR2 BCL2L13 BID LINC00528 MICAL3 MIR3198-1 ATP6V1E1 LOC101929372 SLC25A18 |
dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain | BCL2L13 ATP6V1E1 LOC101929372 SLC25A18 |
esv3647226 | Chr.22:17584572 - 17586555 on Build GRCh38 | Loss | LOC101929372 SLC25A18 |
nsv510802 | Chr.22:17550100 - 17604891 on Build GRCh38 | Deletion | CECR2 ATP6V1E1 LOC101929372 SLC25A18 |
esv2657458 | Chr.22:17583664 - 17586671 on Build GRCh38 | Deletion | LOC101929372 SLC25A18 |
esv3647225 | Chr.22:17583610 - 17586518 on Build GRCh38 | Loss | LOC101929372 SLC25A18 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |