Assay Details
Target Gene Details
Entrez Gene ID: | 27349 |
Gene Name: | malonyl-CoA-acyl carrier protein transacylase |
Gene Aliases: |
FASN2C, MCT, MT, NET62, fabD |
Location: |
Chr.22:43132206-43143397 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MCAT | NM_014507.3 | NP_055322.1 | ||
NM_173467.4 | NP_775738.3 | |||
NR_046423.1 | ||||
XM_011530156.2 | XP_011528458.1 | |||
AA402283.1 | ||||
AK314059.1 | ||||
AL359401.1 | CAB94789.1 | |||
AL359403.1 | CAB94790.1 | |||
BC030985.1 | AAH30985.1 | |||
BC042195.1 | AAH42195.2 | |||
BI464466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762124 | Chr.22:43031130 - 43188670 on Build GRCh38 | Gain | MCAT BIK TTLL1 TSPO LOC100506679 TTLL12 |
esv33816 | Chr.22:43113758 - 43326122 on Build GRCh38 | Gain | MCAT BIK LOC101927447 SCUBE1 TSPO LOC105373051 TTLL12 |
nsv834214 | Chr.22:43037931 - 43217928 on Build GRCh38 | Gain | MCAT BIK TTLL1 SCUBE1 TSPO LOC100506679 LOC105373051 TTLL12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |