Assay Details
Target Gene Details
Entrez Gene ID: | 57553 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 3 |
Gene Aliases: |
MICAL-3 |
Location: |
Chr.22:17787649-18024559 on Build GRCh38 |
Assay Gene Location: | Within Intron 47 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | MIR3198-1 MICAL3 BID LOC101929372 SLC25A18 CECR2 BCL2L13 ATP6V1E1 LINC00528 |
nsv834120 | Chr.22:17733890 - 17904438 on Build GRCh38 | Loss | MIR3198-1 MICAL3 BID LINC00528 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | MIR648 MIR3198-1 MICAL3 FLJ41941 BID LOC101929372 SLC25A18 BCL2L13 ATP6V1E1 LINC00528 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC100996415 MICAL3 LOC100996401 SLC25A18 RIMBP3 MIR3198-1 LOC102725072 PI4KAP1 BCL2L13 ATP6V1E1 LINC00528 LOC729461 TUBA8 LOC642643 BID LOC101929372 TMEM191B CECR2 GGT3P LOC100996432 MIR648 USP18 FAM230A FLJ41941 PEX26 |
nsv834119 | Chr.22:17665133 - 17795994 on Build GRCh38 | Loss | MIR3198-1 MICAL3 BID BCL2L13 LINC00528 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |