Assay Details
Target Gene Details
Entrez Gene ID: | 23764 |
Gene Name: | MAF bZIP transcription factor F |
Gene Aliases: |
U-MAF, hMafF |
Location: |
Chr.22:38201932-38216511 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAFF | NM_001161572.1 | NP_001155044.1 | ||
NM_001161573.1 | NP_001155045.1 | |||
NM_001161574.1 | NP_001155046.1 | |||
NM_012323.3 | NP_036455.1 | |||
AB025247.1 | BAA86871.1 | |||
AJ010857.1 | CAB52435.1 | |||
AK002001.1 | BAA92029.1 | |||
AK300932.1 | ||||
BC015037.1 | AAH15037.1 | |||
BC067751.1 | AAH67751.1 | |||
DA209378.1 | ||||
DA425613.1 | ||||
DA654696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv989237 | Chr.22:38201294 - 38213896 on Build GRCh38 | Deletion | MAFF |
nsv519015 | Chr.22:38172999 - 38210773 on Build GRCh38 | Loss | MAFF PLA2G6 |
nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss | BAIAP2L2 TMEM184B MAFF SLC16A8 SNORA92 PICK1 PLA2G6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |