Assay Details
Target Gene Details
Entrez Gene ID: | 10766 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
APRO5, TOB4, TOBL, TROB2 |
Location: |
Chr.22:41433488-41448230 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB2 | NM_016272.3 | NP_057356.1 | ||
XM_005261315.2 | XP_005261372.1 | |||
XM_006724105.3 | XP_006724168.1 | |||
XM_017028539.1 | XP_016884028.1 | |||
AB051450.1 | ||||
BC028919.1 | AAH28919.1 | |||
BC038957.1 | AAH38957.1 | |||
BM453650.1 | ||||
CR456594.1 | CAG30480.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss | RANGAP1 TOB2 TEF PHF5A LOC100996598 ZC3H7B |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss | MIR6889 ACO2 RANGAP1 LOC105373044 TOB2 TEF PHF5A LOC100996598 POLR3H CSDC2 ZC3H7B PMM1 |
nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss | ACO2 LOC105373044 TOB2 TEF PHF5A LOC100996598 POLR3H |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain | MIR6889 RANGAP1 TOB2 TEF LOC100996598 CHADL ZC3H7B |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss | C22orf46 RANGAP1 LOC105373044 TOB2 XRCC6 MIR1281 PHF5A SNORD140 CSDC2 ZC3H7B MIR6889 MEI1 L3MBTL2 EP300 ACO2 SNU13 EP300-AS1 TEF LOC100996598 POLR3H CHADL DESI1 PMM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |