Assay Details
Target Gene Details
Entrez Gene ID: | 91464 |
Gene Name: | intestine specific homeobox |
Gene Aliases: |
Pix-1, RAXLX |
Location: |
Chr.22:35066136-35087387 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ISX | NM_001303508.1 | NP_001290437.1 | ||
AB219123.1 | ||||
AK025181.1 | ||||
BC112212.1 | AAI12213.1 | |||
BC113633.1 | ||||
CR749372.1 | CAH18225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834182 | Chr.22:34986155 - 35159250 on Build GRCh38 | Gain | ISX-AS1 LINC01399 ISX |
nsv817989 | Chr.22:35066656 - 35154238 on Build GRCh38 | Loss | LINC01399 ISX |
nsv834183 | Chr.22:35048670 - 35233532 on Build GRCh38 | Gain | LINC01399 ISX |
esv35003 | Chr.22:35060310 - 35175504 on Build GRCh38 | Loss | LINC01399 ISX |
esv26909 | Chr.22:35061958 - 35073684 on Build GRCh38 | Gain | ISX |
esv34121 | Chr.22:34988871 - 35101201 on Build GRCh38 | Loss | ISX-AS1 ISX |
esv3647659 | Chr.22:35064756 - 35171556 on Build GRCh38 | Loss | LINC01399 ISX |
esv2661366 | Chr.22:35064758 - 35171614 on Build GRCh38 | Deletion | LINC01399 ISX |
esv2758835 | Chr.22:35042419 - 35236960 on Build GRCh38 | Loss | LINC01399 ISX |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |