Assay Details
Target Gene Details
Entrez Gene ID: | 26150 |
Gene Name: | RIB43A domain with coiled-coils 2 |
Gene Aliases: |
C22orf11, TRIB |
Location: |
Chr.22:45413691-45432422 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RIBC2 | NM_015653.4 | NP_056468.3 | ||
XM_005261524.4 | XP_005261581.1 | |||
XM_011530126.2 | XP_011528428.1 | |||
AK098586.1 | ||||
AK225722.1 | ||||
AL442116.1 | CAC09526.1 | |||
BC003024.1 | AAH03024.1 | |||
BC009904.2 | AAH09904.1 | |||
CR456438.1 | CAG30324.1 | |||
CR457398.1 | CAG33679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3647932 | Chr.22:45418035 - 45484463 on Build GRCh38 | Gain | RIBC2 |
nsv1066204 | Chr.22:45254728 - 45870451 on Build GRCh38 | Gain | UPK3A LOC105373065 MIR4762 LINC01589 FBLN1 ATXN10 SMC1B RIBC2 FAM118A |
nsv3651 | Chr.22:45403708 - 45435552 on Build GRCh38 | Insertion | SMC1B RIBC2 |
nsv834221 | Chr.22:45383909 - 45578311 on Build GRCh38 | Loss | FBLN1 SMC1B RIBC2 |
esv3647931 | Chr.22:45382351 - 45517601 on Build GRCh38 | Gain | FBLN1 SMC1B RIBC2 |
esv34218 | Chr.22:45254944 - 45515710 on Build GRCh38 | Gain | UPK3A LOC105373065 FBLN1 SMC1B RIBC2 FAM118A |
esv2758843 | Chr.22:45178343 - 45529286 on Build GRCh38 | Gain | UPK3A KIAA0930 LOC105373065 NUP50 MIR1249 FBLN1 SMC1B RIBC2 FAM118A |
nsv9921 | Chr.22:45247876 - 45514132 on Build GRCh38 | Gain+Loss | UPK3A LOC105373065 FBLN1 SMC1B RIBC2 FAM118A |
esv3647921 | Chr.22:45247581 - 45517162 on Build GRCh38 | Gain | UPK3A LOC105373065 FBLN1 SMC1B RIBC2 FAM118A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |