Assay Details
Target Gene Details
Entrez Gene ID: | 100874530 |
Gene Name: | APOBEC3B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:38991559-38998220 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBEC3B-AS1 | NR_104187.1 | |||
CX754295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3627 | Chr.22:38952080 - 39000579 on Build GRCh38 | Deletion | APOBEC3B-AS1 APOBEC3A APOBEC3B |
dgv4571n100 | Chr.22:38967602 - 39003295 on Build GRCh38 | Loss | APOBEC3B-AS1 APOBEC3B |
esv2760711 | Chr.22:38955461 - 39003307 on Build GRCh38 | Loss | APOBEC3B-AS1 APOBEC3A APOBEC3B |
dgv172n111 | Chr.22:38947911 - 39007789 on Build GRCh38 | Deletion | APOBEC3B-AS1 APOBEC3A APOBEC3B |
nsv198 | Chr.22:38954302 - 39000579 on Build GRCh38 | Deletion | APOBEC3B-AS1 APOBEC3A APOBEC3B |
nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss | APOBEC3B-AS1 APOBEC3G APOBEC3D APOBEC3F CBX7 APOBEC3C APOBEC3H |