Assay Details
Target Gene Details
Entrez Gene ID: | 9814 |
Gene Name: | SFI1 centrin binding protein |
Gene Aliases: |
PISD, PPP1R139, hSfi1p |
Location: |
Chr.22:31496139-31618554 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3594 | Chr.22:31504103 - 31537708 on Build GRCh38 | Insertion | SFI1 |
nsv1059314 | Chr.22:31497627 - 31540102 on Build GRCh38 | Gain | SFI1 |
nsv588894 | Chr.22:31437773 - 31581608 on Build GRCh38 | Loss | EIF4ENIF1 SFI1 |
nsv1063634 | Chr.22:31401018 - 31525268 on Build GRCh38 | Gain | EIF4ENIF1 DRG1 SFI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |