Assay Details
Target Gene Details
Entrez Gene ID: | 64699 |
Gene Name: | transmembrane protease, serine 3 |
Gene Aliases: |
DFNB10, DFNB8, ECHOS1, TADG12 |
Location: |
Chr.21:42371887-42396846 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS3 | NM_001256317.1 | NP_001243246.1 | ||
NM_024022.2 | NP_076927.1 | |||
NM_032405.1 | NP_115781.1 | |||
NR_046020.1 | ||||
AB038157.1 | BAB20077.1 | |||
AB038160.1 | BAB20080.1 | |||
AF201380.1 | AAG37012.1 | |||
AF267741.1 | AAL56664.1 | |||
AK172842.1 | BAD18806.1 | |||
AY358458.1 | AAQ88823.1 | |||
AY633572.1 | AAT66641.1 | |||
BC074846.2 | AAH74846.1 | |||
BC074847.2 | AAH74847.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |