Assay Details
Target Gene Details
Entrez Gene ID: | 56894 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 3 |
Gene Aliases: |
LPAAT-GAMMA1, LPAAT3 |
Location: |
Chr.21:43865160-43987594 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGPAT3 | NM_020132.4 | NP_064517.1 | ||
XM_006724031.3 | XP_006724094.1 | |||
XM_011529665.2 | XP_011527967.1 | |||
XM_017028409.1 | XP_016883898.1 | |||
XM_017028412.1 | XP_016883901.1 | |||
XM_017028413.1 | XP_016883902.1 | |||
BC011971.1 | AAH11971.1 | |||
CD672915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834108 | Chr.21:43758156 - 43949760 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv33817 | Chr.21:43749531 - 44647625 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)