Assay Details
Target Gene Details
Entrez Gene ID: | 10732 |
Gene Name: | transcription factor like 5 |
Gene Aliases: |
CHA, E2BP-1, Figlb, bHLHe82 |
Location: |
Chr.20:62841014-62861763 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TCFL5 | NM_006602.3 | NP_006593.2 | ||
XM_005260185.3 | XP_005260242.1 | |||
XM_011528497.2 | XP_011526799.1 | |||
XM_011528499.1 | XP_011526801.1 | |||
AB012124.1 | BAA36557.1 | |||
AF070992.1 | AAD53986.1 | |||
AJ271337.1 | CAC24700.1 | |||
BC046933.1 | AAH46933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834027 | Chr.20:62707655 - 62874914 on Build GRCh38 | Loss | LOC107987285 MRGBP COL9A3 NTSR1 DPH3P1 TCFL5 LINC00659 OGFR-AS1 LOC107985428 OGFR |
nsv828794 | Chr.20:62832958 - 62880035 on Build GRCh38 | Loss | COL9A3 DPH3P1 TCFL5 DIDO1 |
nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain | LOC107985425 MIR3196 LOC102723788 NKAIN4 EEF1A2 MIR4326 LINC01056 PTK6 YTHDF1 LINC00659 LOC107985428 MIR124-3 LINC00029 MRGBP ARFGAP1 HAR1B DPH3P1 TCFL5 HELZ2 SRMS CHRNA4 GMEB2 GID8 COL9A3 HAR1A LOC100130587 PPDPF BIRC7 COL20A1 LOC63930 FLJ16779 LOC100130152 KCNQ2 DIDO1 BHLHE23 FNDC11 OGFR-AS1 SLC17A9 OGFR |
nsv428380 | Chr.20:62632018 - 62935383 on Build GRCh38 | Loss | LOC107987285 SLCO4A1 COL9A3 LINC00659 LOC107985428 SLCO4A1-AS1 MRGBP NTSR1 DPH3P1 TCFL5 DIDO1 OGFR-AS1 OGFR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |