Assay Details
Target Gene Details
Entrez Gene ID: | 6939 |
Gene Name: | transcription factor 15 (basic helix-loop-helix) |
Gene Aliases: |
EC2, PARAXIS, bHLHa40 |
Location: |
Chr.20:603993-610266 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TCF15 | NM_004609.3 | NP_004600.2 | ||
AI218364.1 | ||||
AW290965.1 | ||||
BC067836.1 | AAH67836.1 | |||
U08336.1 | AAA19969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7422n54 | Chr.20:606311 - 613017 on Build GRCh38 | Loss | TCF15 |
nsv585173 | Chr.20:592392 - 625245 on Build GRCh38 | Loss | TCF15 |
nsv469833 | Chr.20:447851 - 614798 on Build GRCh38 | Gain | CSNK2A1 TCF15 TBC1D20 |
esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain | CSNK2A1 LOC105372493 RBCK1 TCF15 SRXN1 PSMF1 SLC52A3 RSPO4 TRIB3 SCRT2 ANGPT4 FAM110A TBC1D20 |
nsv458845 | Chr.20:602943 - 625293 on Build GRCh38 | Loss | TCF15 |
nsv833891 | Chr.20:468248 - 620062 on Build GRCh38 | Gain+Loss | CSNK2A1 TCF15 |
nsv828661 | Chr.20:606973 - 612340 on Build GRCh38 | Gain | TCF15 |
nsv458844 | Chr.20:599013 - 614136 on Build GRCh38 | Loss | TCF15 |
nsv585172 | Chr.20:551334 - 613017 on Build GRCh38 | Gain | TCF15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |