Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | NP_001139528.1 | ||
NM_001146082.2 | NP_001139554.1 | |||
NM_001146083.2 | NP_001139555.1 | |||
NM_024298.4 | NP_077274.3 | |||
XM_011527299.2 | XP_011525601.1 | |||
XM_011527300.2 | XP_011525602.1 | |||
XM_017027296.1 | XP_016882785.1 | |||
AK055908.1 | BAB71043.1 | |||
AK122789.1 | ||||
AK293199.1 | ||||
AK298689.1 | ||||
AK311493.1 | ||||
BC002512.2 | AAH02512.2 | |||
BC003164.1 | AAH03164.2 | |||
BC006309.2 | AAH06309.1 | |||
BC015857.1 | ||||
EU016381.1 | ||||
S82470.1 | AAB37433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066425 | Chr.19:54182441 - 54237431 on Build GRCh38 | Gain | MBOAT7 RPS9 LILRA6 TSEN34 LILRB3 |
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | MBOAT7 TMC4 LENG1 TSEN34 |
nsv2546 | Chr.19:54182614 - 54270671 on Build GRCh38 | Deletion | MBOAT7 RPS9 LILRA6 TSEN34 LILRB3 LILRB5 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | MBOAT7 CNOT3 NDUFA3 TMC4 LENG1 TSEN34 OSCAR TFPT PRPF31 |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | MIR519A2 ZNF813 MIR517A MIR526B LENG1 BIRC8 MIR527 TFPT MIR935 LILRB3 MIR519B CACNG7 NDUFA3 MIR517B ZNF845 ZNF818P MIR512-2 TARM1 MIR520D MIR516A1 MIR521-2 MIR524 TPM3P9 MIR520A MIR518C MIR523 MIR526A2 MIR518F MIR519A1 MIR520B ZNF677 LILRB5 CACNG6 MIR520G LILRA6 MIR520C LILRA5 MIR522 MIR518B MIR526A1 LOC284379 ZNF761 MIR516B1 MIR519E PRPF31 PRKCG MIR1283-2 MIR512-1 MIR519D VN1R2 FAM90A27P RPS9 VN1R4 MIR4752 MIR516A2 NLRP12 TSEN34 DPRX MIR518A2 MIR518D ZNF665 CNOT3 ZNF525 TMC4 MIR372 LILRB2 CACNG8 MIR1283-1 MIR525 MIR371A MIR515-1 MIR520H MIR498 ZNF765 MIR520E MIR515-2 LOC105372457 MIR371B MIR1323 MIR518E VSTM1 MYADM MIR521-1 MBOAT7 MIR518A1 MIR520F ZNF331 MIR519C MIR373 MIR517C OSCAR MIR516B2 |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | MBOAT7 RPS9 LILRA6 TMC4 LENG1 TSEN34 LILRB3 LILRB5 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | LENG8 RPS9 MIR4752 KIR3DL1 TSEN34 TTYH1 LENG9 NCR1 LILRB3 LILRA4 LILRB4 LOC101928804 LILRA1 TMC4 LILRB2 KIR3DL3 LAIR1 LOC102724300 KIR2DL3 LILRB1 KIR3DL2 KIR2DL1 LILRA2 LILRB5 LILRP2 LAIR2 KIR2DS4 KIR3DX1 MBOAT7 LILRA6 LENG8-AS1 KIR2DL4 LILRA5 CDC42EP5 FCAR MIR8061 NLRP7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |