Assay Details
Target Gene Details
Entrez Gene ID: | 54776 |
Gene Name: | protein phosphatase 1 regulatory subunit 12C |
Gene Aliases: |
LENG3, MBS85, p84, p85 |
Location: |
Chr.19:55090913-55117600 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R12C | NM_001271618.1 | NP_001258547.1 | ||
NM_017607.3 | NP_060077.1 | |||
XM_005259013.3 | XP_005259070.1 | |||
XM_011527045.2 | XP_011525347.1 | |||
AB209452.1 | BAD92689.1 | |||
AF312028.1 | AAG60045.1 | |||
AK027086.1 | BAB15651.1 | |||
AK297426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055961 | Chr.19:55102359 - 55132708 on Build GRCh38 | Gain+Loss | PPP1R12C MIR7975 TNNT1 |
esv3644836 | Chr.19:55084362 - 55113486 on Build GRCh38 | Gain | EPS8L1 PPP1R12C |
esv3644829 | Chr.19:55030893 - 55138732 on Build GRCh38 | Gain | EPS8L1 RDH13 PPP1R12C GP6 MIR7975 TNNT1 |
esv2758772 | Chr.19:55026094 - 55139319 on Build GRCh38 | Gain | EPS8L1 RDH13 PPP1R12C GP6 MIR7975 TNNT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |