Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
AK124343.1 | ||||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SNAR-A2 PNKP MED25 HSD17B14 TSKS CGB2 PLA2G4C ALDH16A1 RRAS SNAR-A10 SNORD32A LOC100287477 NTN5 KCNJ14 BAX PPFIA3 AP2A1 SNAR-B2 TEAD2 SYNGR4 CGB5 NUP62 RPL13A TULP2 BCL2L12 RASIP1 SNORD34 NUCB1 SNAR-A9 SNAR-A1 FUT2 ZNF473 NUCB1-AS1 PTOV1-AS1 CCDC114 SNAR-A6 RPS11 SLC17A7 FTL SNAR-C2 MIR6800 CGB3 PRMT1 SNAR-A13 AKT1S1 FGF21 PLEKHA4 KDELR1 SIGLEC11 TBC1D17 LOC101928295 SLC6A16 CARD8 SNAR-B1 SNORD35A CCDC155 LHB SNAR-A3 SEC1P GYS1 CGB7 NTF4 RPL18 SIGLEC16 LMTK3 CA11 PTOV1 KCNA7 GRIN2D SNAR-A14 SNAR-A5 IL4I1 SNRNP70 LIN7B CYTH2 FLT3LG LIG1 MIR4751 PRR12 PLA2G4C-AS1 PPP1R15A SNAR-C3 ATF5 LOC105447645 FUZ CPT1C SCAF1 LOC101059948 SNAR-A4 CGB8 MIR6799 BSPH1 SNAR-A8 MIR4749 GFY SNAR-C5 SNORD35B IZUMO1 MIR4750 FAM83E SNAR-C4 SPHK2 TMEM143 LOC105372430 SNAR-G2 C19orf73 MAMSTR SNAR-C1 CD37 NOSIP PTOV1-AS2 TRPM4 RUVBL2 ZNF114 DKKL1 IRF3 FCGRT SPACA4 PTH2 HRC BCAT2 FLJ26850 SULT2B1 SNAR-G1 ADM5 MIR5088 DBP CGB1 SNAR-A12 MIR6798 SNAR-A7 SNORD33 EMP3 C19orf68 PIH1D1 VRK3 PRRG2 MIR150 FUT1 ELSPBP1 SNAR-D CABP5 RCN3 CARD8-AS1 GRWD1 DHDH SNAR-A11 MIR4324 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | BCL2L12 PTOV1-AS1 PRR12 MED25 FUZ TSKS PTOV1 MIR6800 CPT1C SCAF1 MIR6799 RRAS PRMT1 ADM5 AP2A1 MIR4749 PTOV1-AS2 MIR5088 IRF3 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | PNKP MIR4750 MIR4751 PTOV1-AS1 MED25 ATF5 FUZ TSKS PTOV1 MIR6800 MIR6799 IL4I1 AKT1S1 AP2A1 MIR4749 TBC1D17 PTOV1-AS2 NUP62 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | PNKP MIR4751 PTOV1-AS1 PRR12 MED25 ATF5 FUZ TSKS MIR6800 CPT1C SCAF1 MIR6799 RRAS PRMT1 AKT1S1 ADM5 AP2A1 MIR4749 TBC1D17 MIR5088 NUP62 MIR4750 BCL2L12 PTOV1 IL4I1 PRRG2 NOSIP PTOV1-AS2 IRF3 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | PTOV1-AS1 PRR12 MED25 RPS11 FUZ TSKS MIR6800 CPT1C SCAF1 MIR6799 RRAS PRMT1 SNORD32A ADM5 AP2A1 SNORD35B MIR5088 SNORD35A RPL13A BCL2L12 SNORD34 SNORD33 PRRG2 MIR150 NOSIP RCN3 IRF3 FCGRT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |