Assay Details
Target Gene Details
Entrez Gene ID: | 1954 |
Gene Name: | multiple EGF like domains 8 |
Gene Aliases: |
C19orf49, CRPT2, EGFL4, SBP1 |
Location: |
Chr.19:42325609-42378769 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEGF8 | NM_001271938.1 | NP_001258867.1 | ||
NM_001410.2 | NP_001401.2 | |||
AB011541.2 | BAA32469.2 | |||
BC153880.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | ATP1A3 ZNF574 LOC100505622 ZNF526 MIR6797 MIR4323 POU2F2 CNFN DEDD2 RPS19 ERF LOC390937 TMEM145 LOC100505585 RABAC1 CIC GSK3A ARHGEF1 MEGF8 LIPE-AS1 PAFAH1B3 LOC101930071 GRIK5 CD79A MIR8077 PRR19 LIPE |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | TMEM145 CIC CEACAM1 GSK3A CEACAM8 ZNF526 MEGF8 LIPE-AS1 PAFAH1B3 LOC101930071 POU2F2 CNFN DEDD2 CXCL17 MIR8077 PRR19 LIPE ERF |
nsv1160627 | Chr.19:42209979 - 42341899 on Build GRCh38 | Deletion | TMEM145 CIC DEDD2 GSK3A ZNF526 PRR19 MEGF8 PAFAH1B3 ERF |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | ATP1A3 LYPD4 DMRTC2 ZNF574 LOC100505622 ZNF526 CEACAM5 MIR6797 MIR4323 POU2F2 CNFN DEDD2 RPS19 ERF LOC390937 TMEM145 LOC100505585 RABAC1 CIC CEACAM7 GSK3A CEACAM6 ARHGEF1 MEGF8 LIPE-AS1 PAFAH1B3 LOC101930071 GRIK5 CD79A MIR8077 PRR19 LIPE CEACAM3 CEACAM4 |
nsv833834 | Chr.19:42232292 - 42330875 on Build GRCh38 | Loss | TMEM145 CIC GSK3A PRR19 MEGF8 PAFAH1B3 ERF |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | TMEM145 PSG11 CIC PSG2 CEACAM1 GSK3A CEACAM8 MEGF8 LIPE-AS1 PSG3 PSG6 PAFAH1B3 LOC101930071 PSG10P CNFN PSG1 PSG7 CXCL17 PSG8 LOC100289650 MIR8077 PRR19 LIPE ERF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |