Assay Details
Target Gene Details
Entrez Gene ID: | 51599 |
Gene Name: | lipolysis stimulated lipoprotein receptor |
Gene Aliases: |
ILDR3, LISCH7 |
Location: |
Chr.19:35248656-35267964 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LSR | NM_001260489.1 | NP_001247418.1 | ||
NM_001260490.1 | NP_001247419.1 | |||
NM_015925.6 | NP_057009.3 | |||
NM_205834.3 | NP_991403.1 | |||
NM_205835.3 | NP_991404.1 | |||
XM_005258980.1 | XP_005259037.1 | |||
XM_005258982.1 | XP_005259039.1 | |||
XM_011527026.1 | XP_011525328.1 | |||
AF130366.1 | AAD22101.1 | |||
AK075426.1 | BAC11614.1 | |||
AK126834.1 | BAC86714.1 | |||
AK130367.1 | ||||
AK296618.1 | ||||
BC000015.2 | AAH00015.2 | |||
BC004381.1 | ||||
BC047376.1 | AAH47376.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2665499 | Chr.19:35253070 - 35263082 on Build GRCh38 | Deletion | LSR |
esv2758758 | Chr.19:35220175 - 35396211 on Build GRCh38 | Loss | LSR FAM187B USF2 MAG FFAR1 HAMP MIR5196 FFAR3 CD22 |
esv3644232 | Chr.19:35253087 - 35263050 on Build GRCh38 | Loss | LSR |
nsv953283 | Chr.19:35247998 - 35314097 on Build GRCh38 | Deletion | LSR USF2 MAG HAMP |
nsv510765 | Chr.19:35217323 - 35399766 on Build GRCh38 | Deletion | LSR FAM187B USF2 MAG FFAR1 HAMP MIR5196 FFAR3 CD22 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |