Assay Details
Target Gene Details
Entrez Gene ID: | 100529262 |
Gene Name: | MIA-RAB4B readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.19:40775537-40796944 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIA-RAB4B | NR_037775.1 | |||
AF087861.1 | AAP97171.1 |
Target Gene Details
Entrez Gene ID: | 53916 |
Gene Name: | RAB4B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.19:40778219-40796944 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB4B | NM_016154.4 | NP_057238.3 | ||
AF165522.1 | AAD45923.1 | |||
AF217985.1 | AAG17228.1 | |||
AI083750.1 | ||||
AK024845.1 | ||||
BC033081.1 | AAH33081.1 | |||
BC046927.1 | AAH46927.1 |
Target Gene Details
Entrez Gene ID: | 100529264 |
Gene Name: | RAB4B-EGLN2 readthrough (NMD candidate) |
Gene Aliases: |
RERT-lncRNA |
Location: |
Chr.19:40778219-40808441 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB4B-EGLN2 | NR_037791.1 | |||
AK291385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833832 | Chr.19:40754458 - 40828692 on Build GRCh38 | Loss | RAB4B-EGLN2 MIA CYP2T1P RAB4B MIA-RAB4B EGLN2 SNRPA |
esv2758762 | Chr.19:40785722 - 41040163 on Build GRCh38 | Gain+Loss | RAB4B-EGLN2 CYP2B7P CYP2A7P1 CYP2T1P CYP2A6 RAB4B CYP2A7 CYP2G1P MIA-RAB4B EGLN2 CYP2B6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |