Assay Details
Target Gene Details
Entrez Gene ID: | 1954 |
Gene Name: | multiple EGF like domains 8 |
Gene Aliases: |
C19orf49, CRPT2, EGFL4, SBP1 |
Location: |
Chr.19:42325609-42378769 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEGF8 | NM_001271938.1 | NP_001258867.1 | ||
NM_001410.2 | NP_001401.2 | |||
AB011541.2 | BAA32469.2 | |||
BC153880.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | RABAC1 GRIK5 ARHGEF1 ZNF526 MIR6797 LIPE LIPE-AS1 CNFN LOC100505585 PAFAH1B3 GSK3A PRR19 CD79A LOC101930071 LOC390937 ZNF574 MIR4323 CIC TMEM145 MEGF8 POU2F2 LOC100505622 ERF MIR8077 RPS19 ATP1A3 DEDD2 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | LOC101930071 CIC ZNF526 TMEM145 LIPE CEACAM8 MEGF8 POU2F2 ERF LIPE-AS1 CNFN CEACAM1 MIR8077 PAFAH1B3 DEDD2 CXCL17 GSK3A PRR19 |
nsv1160627 | Chr.19:42209979 - 42341899 on Build GRCh38 | Deletion | MEGF8 ERF CIC PAFAH1B3 ZNF526 DEDD2 GSK3A TMEM145 PRR19 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | DMRTC2 RABAC1 GRIK5 CEACAM4 ARHGEF1 ZNF526 MIR6797 LIPE CEACAM5 LIPE-AS1 CNFN LOC100505585 PAFAH1B3 GSK3A PRR19 CD79A LOC101930071 LOC390937 LYPD4 ZNF574 CEACAM7 MIR4323 CIC TMEM145 CEACAM6 MEGF8 POU2F2 LOC100505622 ERF MIR8077 RPS19 ATP1A3 DEDD2 CEACAM3 |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | LOC101930071 PSG10P LOC100289650 PSG3 CIC PSG8 TMEM145 PSG6 LIPE CEACAM8 PSG11 MEGF8 ERF LIPE-AS1 CNFN CEACAM1 PSG2 MIR8077 PAFAH1B3 CXCL17 PSG7 GSK3A PRR19 PSG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |