Assay Details
Target Gene Details
Entrez Gene ID: | 2208 |
Gene Name: | Fc fragment of IgE receptor II |
Gene Aliases: |
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF |
Location: |
Chr.19:7688757-7702755 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCER2 | NM_001220500.1 | NP_001207429.1 | ||
NM_002002.4 | NP_001993.2 | |||
XM_005272462.4 | XP_005272519.1 | |||
AK223207.1 | BAD96927.1 | |||
AK310071.1 | ||||
AK310707.1 | ||||
BC014108.2 | AAH14108.1 | |||
BC062591.1 | AAH62591.1 | |||
DA957269.1 | ||||
M14766.1 | AAA52435.1 | |||
M15059.1 | AAA52434.1 | |||
X04772.1 | CAA28465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953959 | Chr.19:7669315 - 7700414 on Build GRCh38 | Deletion | TRAPPC5 MCEMP1 FCER2 RETN |
nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss | TRAPPC5 CLEC4G PCP2 MCEMP1 PET100 FCER2 CAMSAP3 MIR6792 XAB2 STXBP2 RETN |
nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain | INSR TRAPPC5 ARHGEF18 PET100 PNPLA6 CAMSAP3 MIR6792 CLEC4M STXBP2 CD209 ZNF358 EVI5L CLEC4G LOC100128573 PCP2 CLEC4GP1 LOC107985283 MCEMP1 PEX11G MCOLN1 FCER2 C19orf45 XAB2 RETN |
nsv833732 | Chr.19:7672656 - 7871329 on Build GRCh38 | Gain | TRAPPC5 EVI5L CLEC4G PRR36 CLEC4GP1 MCEMP1 FCER2 CLEC4M CD209 |
esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain | TRAPPC5 MBD3L2 ARHGEF18 PET100 MIR6792 STXBP2 CD209 MBD3L5 PCP2 LOC107985283 PEX11G FCER2 ADGRE1 MBD3L4 XAB2 INSR PNPLA6 ADGRE4P MBD3L3 CAMSAP3 CLEC4M FLJ25758 LOC729458 ZNF358 CLEC4G LOC100128573 ZNF557 CLEC4GP1 MCEMP1 MCOLN1 C19orf45 RETN |
nsv1061452 | Chr.19:7657750 - 7785131 on Build GRCh38 | Gain | TRAPPC5 CLEC4G MCEMP1 FCER2 CLEC4M CD209 RETN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |