Assay Details
Target Gene Details
Entrez Gene ID: | 100506033 |
Gene Name: | PTOV1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:49838645-49851676 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1-AS1 | NR_040037.1 | |||
AK092213.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SNAR-G1 LOC101928295 CGB2 SNAR-A7 RCN3 MIR4751 SLC17A7 LOC101059948 SPHK2 SYNGR4 SNAR-A12 RASIP1 RPL18 HSD17B14 LHB CGB1 KCNA7 FCGRT GRIN2D SNAR-A1 NUCB1-AS1 SNAR-A2 C19orf73 FGF21 TULP2 SNAR-A10 BCAT2 BAX ADM5 C19orf68 LOC105447645 PNKP BCL2L12 AKT1S1 SIGLEC16 SNRNP70 LOC105372430 CARD8-AS1 CGB3 MIR5088 CPT1C NOSIP FLT3LG MIR6799 SNAR-D RRAS PPP1R15A HRC SNAR-C4 TSKS KCNJ14 ATF5 CABP5 LOC100287477 CGB8 LIN7B SNAR-A14 TEAD2 SNORD34 RPL13A SNAR-A5 SNAR-B2 BSPH1 PLEKHA4 PTOV1-AS1 SNAR-A4 NUCB1 SNAR-A13 SNAR-A9 PTOV1-AS2 CCDC114 GRWD1 CD37 SNORD35B CARD8 SNAR-C5 SNAR-A8 PLA2G4C LMTK3 ALDH16A1 DHDH PPFIA3 KDELR1 SULT2B1 PRR12 SCAF1 EMP3 PLA2G4C-AS1 FUZ ZNF114 SNORD33 SNAR-A3 MIR150 SPACA4 FLJ26850 MAMSTR GYS1 SIGLEC11 SNAR-G2 SEC1P DBP CGB5 DKKL1 ELSPBP1 MIR4749 IL4I1 FUT2 RPS11 LIG1 MIR4324 SNAR-C2 CA11 SNAR-B1 SNORD32A FUT1 CCDC155 PIH1D1 TMEM143 MED25 IRF3 NTN5 ZNF473 VRK3 NUP62 MIR6798 GFY CGB7 RUVBL2 MIR6800 FAM83E SNORD35A TRPM4 SNAR-C3 CYTH2 NTF4 SNAR-A11 SLC6A16 AP2A1 PRMT1 SNAR-C1 IZUMO1 PTOV1 TBC1D17 PRRG2 MIR4750 PTH2 SNAR-A6 FTL |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | AP2A1 PRMT1 BCL2L12 TSKS PRR12 MED25 IRF3 SCAF1 PTOV1-AS1 FUZ PTOV1 PTOV1-AS2 MIR5088 CPT1C MIR6800 MIR4749 MIR6799 ADM5 RRAS |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | AP2A1 PNKP TSKS MED25 AKT1S1 ATF5 PTOV1-AS1 FUZ PTOV1 NUP62 PTOV1-AS2 MIR4751 TBC1D17 MIR4750 MIR6800 MIR4749 IL4I1 MIR6799 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | PNKP BCL2L12 MED25 AKT1S1 IRF3 PTOV1-AS1 NUP62 PTOV1-AS2 MIR4751 MIR5088 CPT1C NOSIP MIR6800 MIR6799 RRAS AP2A1 PRMT1 TSKS PRR12 SCAF1 ATF5 FUZ PTOV1 TBC1D17 PRRG2 MIR4750 MIR4749 IL4I1 ADM5 |