Assay Details
Target Gene Details
Entrez Gene ID: | 2208 |
Gene Name: | Fc fragment of IgE receptor II |
Gene Aliases: |
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF |
Location: |
Chr.19:7688757-7702755 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCER2 | NM_001207019.2 | NP_001193948.2 | ||
NM_001220500.1 | NP_001207429.1 | |||
NM_002002.4 | NP_001993.2 | |||
XM_005272462.4 | XP_005272519.1 | |||
AK223207.1 | BAD96927.1 | |||
AK310071.1 | ||||
BC014108.2 | AAH14108.1 | |||
BC062591.1 | AAH62591.1 | |||
M14766.1 | AAA52435.1 | |||
M15059.1 | AAA52434.1 | |||
X04772.1 | CAA28465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953959 | Chr.19:7669315 - 7700414 on Build GRCh38 | Deletion | FCER2 TRAPPC5 MCEMP1 RETN |
nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss | FCER2 XAB2 CAMSAP3 STXBP2 PCP2 CLEC4G PET100 TRAPPC5 MIR6792 MCEMP1 RETN |
nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain | PEX11G FCER2 XAB2 EVI5L ARHGEF18 LOC100128573 MCOLN1 PET100 INSR MIR6792 MCEMP1 CLEC4GP1 C19orf45 CAMSAP3 CD209 STXBP2 PCP2 CLEC4G ZNF358 PNPLA6 TRAPPC5 CLEC4M LOC107985283 RETN |
nsv578535 | Chr.19:7693308 - 7695738 on Build GRCh38 | Loss | FCER2 |
nsv833732 | Chr.19:7672656 - 7871329 on Build GRCh38 | Gain | FCER2 EVI5L PRR36 CD209 CLEC4G TRAPPC5 CLEC4M MCEMP1 CLEC4GP1 |
esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain | XAB2 MBD3L5 ADGRE4P LOC729458 ADGRE1 MBD3L2 MIR6792 MCEMP1 CLEC4GP1 PCP2 CLEC4G PNPLA6 CLEC4M LOC107985283 PEX11G FCER2 ARHGEF18 LOC100128573 MCOLN1 PET100 FLJ25758 INSR C19orf45 ZNF557 CAMSAP3 CD209 STXBP2 MBD3L3 MBD3L4 ZNF358 TRAPPC5 RETN |
esv26340 | Chr.19:7690078 - 7698176 on Build GRCh38 | Gain | FCER2 |
nsv1061452 | Chr.19:7657750 - 7785131 on Build GRCh38 | Gain | FCER2 CD209 CLEC4G TRAPPC5 CLEC4M MCEMP1 RETN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |