Assay Details
Target Gene Details
Entrez Gene ID: | 2821 |
Gene Name: | glucose-6-phosphate isomerase |
Gene Aliases: |
AMF, GNPI, NLK, PGI, PHI, SA-36, SA36 |
Location: |
Chr.19:34359592-34402413 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPI | XM_011526754.2 | XP_011525056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3644206 | Chr.19:34296741 - 34419464 on Build GRCh38 | Loss | KIAA0355 PDCD2L GPI |
nsv509738 | Chr.19:34351799 - 34412408 on Build GRCh38 | Insertion | KIAA0355 PDCD2L GPI |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |