Assay Details
Target Gene Details
Entrez Gene ID: | 10856 |
Gene Name: | RuvB like AAA ATPase 2 |
Gene Aliases: |
CGI-46, ECP-51, ECP51, INO80J, REPTIN, RVB2, TAP54-beta, TIH2, TIP48, TIP49B |
Location: |
Chr.19:48993448-49015995 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RUVBL2 | NM_001321190.1 | NP_001308119.1 | ||
NM_001321191.1 | NP_001308120.1 | |||
NM_006666.2 | NP_006657.1 | |||
NR_135578.1 | ||||
XM_011526330.1 | XP_011524632.1 | |||
AB024301.1 | BAA76708.1 | |||
AF124607.1 | AAF87087.1 | |||
AF151804.1 | AAD34041.1 | |||
AF155138.1 | AAD38073.1 | |||
AK027762.1 | ||||
AK057498.1 | ||||
AK074542.1 | BAC11048.1 | |||
AK301344.1 | ||||
AL136743.1 | CAB66677.1 | |||
BC000428.2 | AAH00428.1 | |||
BC004531.2 | AAH04531.1 | |||
BC008355.1 | AAH08355.1 | |||
CR533507.1 | CAG38538.1 | |||
Y18417.1 | CAB46270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833859 | Chr.19:48997009 - 49173266 on Build GRCh38 | Loss | CGB5 CGB2 C19orf73 TRPM4 SNAR-G1 HRC CGB7 KCNA7 LHB SNAR-G2 LOC101059948 MIR6798 LIN7B RUVBL2 CGB1 PPFIA3 CGB3 CGB8 SNRNP70 NTF4 |
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SNAR-B1 ALDH16A1 IL4I1 CGB2 ATF5 EMP3 FAM83E BAX PTOV1-AS2 HRC SNAR-A11 CABP5 CYTH2 MIR150 PRRG2 ZNF473 GRIN2D KCNJ14 PPFIA3 SLC6A16 TULP2 NUCB1 SNAR-A13 RPL18 MIR6800 LOC101928295 SNORD35A CGB7 SNORD35B PTH2 LOC101059948 VRK3 RUVBL2 SIGLEC11 BCL2L12 NOSIP CGB8 ELSPBP1 SCAF1 SNAR-A10 CARD8-AS1 MED25 FCGRT BCAT2 LHB PNKP LOC105372430 ADM5 FLJ26850 CPT1C C19orf68 IRF3 LIN7B SNORD32A FTL LOC100287477 SNAR-A7 PRR12 FGF21 PIH1D1 DHDH SNAR-G1 CCDC155 MIR4324 MIR6799 DBP NUCB1-AS1 MIR4751 SNAR-A2 PLEKHA4 MIR5088 SNAR-A3 FUZ DKKL1 SNAR-A9 SPHK2 SIGLEC16 SNAR-C5 AKT1S1 LIG1 LOC105447645 SNAR-C4 FUT1 SNAR-A6 SNAR-A8 SNAR-A1 GYS1 GRWD1 AP2A1 SNAR-A4 FUT2 CGB5 SNAR-A12 CARD8 IZUMO1 TBC1D17 PTOV1-AS1 KDELR1 CA11 LMTK3 PPP1R15A MIR6798 RRAS SNAR-C2 SNAR-D RPS11 NTF4 MIR4750 RPL13A PRMT1 SNAR-A14 GFY SNAR-B2 SLC17A7 TEAD2 FLT3LG SNAR-G2 TSKS CCDC114 SULT2B1 SPACA4 NUP62 PLA2G4C-AS1 RASIP1 SEC1P ZNF114 RCN3 SNAR-C1 C19orf73 CD37 TRPM4 PLA2G4C SNORD34 BSPH1 PTOV1 SNAR-A5 MIR4749 KCNA7 SNAR-C3 NTN5 MAMSTR SYNGR4 CGB1 TMEM143 HSD17B14 CGB3 SNRNP70 SNORD33 |
dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain | FTL LHB GYS1 LOC101059948 MIR6798 RUVBL2 BAX |
nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss | CGB5 FTL NUCB1 CGB2 C19orf73 DHDH BAX SNAR-G1 CGB7 KCNA7 LHB SNAR-G2 GYS1 LOC101059948 MIR6798 NUCB1-AS1 LIN7B RUVBL2 CGB1 PPFIA3 CGB3 CGB8 SNRNP70 NTF4 |
nsv953598 | Chr.19:48957644 - 49085143 on Build GRCh38 | Deletion | CGB5 FTL CGB2 BAX SNAR-G1 CGB7 KCNA7 LHB SNAR-G2 GYS1 LOC101059948 MIR6798 RUVBL2 CGB1 CGB3 CGB8 SNRNP70 NTF4 |
More Information
Additional Information:
For this assay, SNP(s) [rs111856338] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |