Assay Details
Target Gene Details
Entrez Gene ID: | 53635 |
Gene Name: | prostate tumor overexpressed 1 |
Gene Aliases: |
ACID2, PTOV-1 |
Location: |
Chr.19:49850726-49860744 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1 | NM_001305105.1 | NP_001292034.1 | ||
NM_001305108.1 | NP_001292037.1 | |||
NM_017432.4 | NP_059128.2 | |||
NR_130963.1 | ||||
XM_005258998.4 | XP_005259055.1 | |||
XM_006723244.3 | XP_006723307.1 | |||
XM_011527033.1 | XP_011525335.1 | |||
XM_011527034.2 | XP_011525336.1 | |||
XM_011527035.1 | XP_011525337.1 | |||
XM_017026879.1 | XP_016882368.1 | |||
XM_017026880.1 | XP_016882369.1 | |||
AF218026.1 | AAG17268.1 | |||
AJ617480.1 | CAE84582.1 | |||
AK057817.1 | ||||
AY358168.1 | AAQ88535.1 | |||
BC015172.1 | AAH15172.1 | |||
U79287.1 |
Target Gene Details
Entrez Gene ID: | 101928378 |
Gene Name: | PTOV1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.19:49856465-49859289 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1-AS2 | NR_110730.1 | |||
CK903988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | PLA2G4C-AS1 HRC NTN5 GYS1 SNAR-A14 RASIP1 SULT2B1 SNAR-C5 KDELR1 LMTK3 CARD8-AS1 PTOV1-AS1 FTL HSD17B14 CCDC155 SNORD35A PRMT1 FLJ26850 SNAR-G2 ZNF114 SNAR-B1 KCNA7 TSKS SNAR-G1 LIG1 MED25 SNAR-A4 PTH2 PTOV1-AS2 SNAR-D NOSIP SEC1P DKKL1 FUZ GFY IZUMO1 DHDH SNAR-A12 MIR150 LHB LOC105447645 SNAR-A11 RRAS LIN7B FUT2 SNORD33 RPL13A NUCB1-AS1 C19orf73 EMP3 NTF4 CGB7 ATF5 AP2A1 MIR4751 SYNGR4 MIR4750 FLT3LG MIR6799 PNKP MIR6798 CGB5 SNAR-C4 SNAR-A3 AKT1S1 SNAR-A10 IL4I1 MIR4749 SNAR-A1 TBC1D17 RCN3 CABP5 CPT1C CARD8 SNAR-A2 SPHK2 SNAR-C3 PLEKHA4 SIGLEC11 TULP2 CGB2 FAM83E PLA2G4C GRIN2D DBP BSPH1 SNORD34 FGF21 IRF3 SNAR-A13 SNAR-A8 ZNF473 PRRG2 FCGRT PPP1R15A ADM5 TRPM4 CA11 CGB3 SLC6A16 RPS11 PIH1D1 ELSPBP1 TMEM143 PPFIA3 BAX PRR12 CYTH2 SNAR-A9 SNORD35B SIGLEC16 SNORD32A ALDH16A1 GRWD1 SNRNP70 LOC105372430 SNAR-C2 CGB8 MAMSTR SCAF1 SPACA4 TEAD2 C19orf68 SNAR-B2 SNAR-A6 CGB1 LOC101928295 NUP62 RPL18 SLC17A7 LOC100287477 MIR6800 MIR5088 BCAT2 KCNJ14 FUT1 MIR4324 SNAR-A7 CCDC114 VRK3 CD37 LOC101059948 SNAR-C1 RUVBL2 NUCB1 SNAR-A5 PTOV1 BCL2L12 |
nsv526372 | Chr.19:49853907 - 49877601 on Build GRCh38 | Loss | PNKP MIR4749 TBC1D17 PTOV1-AS2 PTOV1 AKT1S1 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | MIR6800 IL4I1 MIR4749 TBC1D17 ATF5 AP2A1 MIR4751 MIR4750 MIR6799 PNKP TSKS MED25 PTOV1-AS1 PTOV1-AS2 PTOV1 NUP62 AKT1S1 FUZ |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | IL4I1 MIR4749 TBC1D17 CPT1C SCAF1 RRAS PTOV1-AS1 IRF3 NUP62 MIR6800 PRRG2 MIR5088 ADM5 PRMT1 ATF5 AP2A1 MIR4751 MIR4750 MIR6799 PNKP TSKS MED25 PTOV1-AS2 PTOV1 PRR12 NOSIP BCL2L12 AKT1S1 FUZ |
dgv54n68 | Chr.19:49848512 - 50008788 on Build GRCh38 | Loss | SIGLEC16 IL4I1 MIR4749 TBC1D17 SIGLEC11 ATF5 MIR4751 MIR4750 PNKP VRK3 PTOV1-AS1 PTOV1-AS2 PTOV1 NUP62 AKT1S1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |