Assay Details
Target Gene Details
Entrez Gene ID: | 100506033 |
Gene Name: | PTOV1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:49838645-49851676 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1-AS1 | NR_040037.1 | |||
AK092213.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | ELSPBP1 RPL18 SNRNP70 FTL CGB5 KCNJ14 SNAR-A6 LOC105372430 SIGLEC11 BAX SLC6A16 DKKL1 PLEKHA4 LIN7B RUVBL2 GYS1 SYNGR4 CARD8-AS1 SULT2B1 SNAR-B1 SNAR-C1 SPACA4 MIR4749 MIR5088 NUCB1-AS1 LOC105447645 MIR6798 MAMSTR FLJ26850 SCAF1 ZNF114 MIR4751 IL4I1 C19orf73 MIR6799 SNAR-C2 SNAR-B2 CCDC155 TMEM143 CD37 SNAR-A11 CGB8 TBC1D17 SNAR-A4 TRPM4 DBP NOSIP C19orf68 HSD17B14 TSKS SNORD34 NUCB1 PRR12 TULP2 GFY FUT2 ATF5 SIGLEC16 LMTK3 SLC17A7 SNORD33 PLA2G4C-AS1 CGB2 NUP62 LOC101059948 SNAR-C4 PRRG2 CGB1 LHB BCAT2 RASIP1 SNAR-G1 MIR4750 KCNA7 SNORD35A PTH2 RPL13A CYTH2 SNAR-A12 SNORD35B CGB3 GRIN2D FAM83E FCGRT PTOV1-AS2 SNAR-G2 ADM5 VRK3 FGF21 SNAR-D BCL2L12 MED25 SNAR-C5 FLT3LG PTOV1-AS1 RCN3 CPT1C SNAR-A9 EMP3 ALDH16A1 HRC MIR4324 KDELR1 MIR150 IZUMO1 RPS11 CGB7 PPFIA3 IRF3 SNAR-C3 TEAD2 PNKP LOC100287477 FUT1 CA11 PIH1D1 SNAR-A7 BSPH1 SEC1P CCDC114 SNAR-A1 FUZ LIG1 SNORD32A CABP5 MIR6800 PPP1R15A SNAR-A3 PLA2G4C SNAR-A8 SNAR-A10 RRAS AKT1S1 AP2A1 SNAR-A5 LOC101928295 CARD8 PRMT1 SNAR-A2 NTN5 SPHK2 NTF4 DHDH GRWD1 ZNF473 SNAR-A13 PTOV1 SNAR-A14 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | PTOV1-AS1 CPT1C RRAS AP2A1 MIR4749 PRMT1 MIR5088 PTOV1-AS2 ADM5 FUZ SCAF1 BCL2L12 TSKS PTOV1 MED25 IRF3 MIR6800 MIR6799 PRR12 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | PTOV1-AS1 MIR4750 PNKP TBC1D17 AKT1S1 ATF5 AP2A1 MIR4749 PTOV1-AS2 NUP62 FUZ TSKS PTOV1 MED25 MIR4751 IL4I1 MIR6800 MIR6799 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | MIR4750 PNKP TBC1D17 NOSIP PTOV1-AS2 ADM5 FUZ BCL2L12 TSKS MED25 MIR6800 PRR12 PTOV1-AS1 CPT1C RRAS AKT1S1 ATF5 AP2A1 MIR4749 PRMT1 MIR5088 NUP62 SCAF1 PRRG2 PTOV1 MIR4751 IL4I1 IRF3 MIR6799 |