Assay Details
Target Gene Details
Entrez Gene ID: | 83855 |
Gene Name: | Kruppel like factor 16 |
Gene Aliases: |
BTEB4, DRRF, NSLP2 |
Location: |
Chr.19:1852399-1863565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF16 | NM_031918.3 | NP_114124.1 | ||
AF327440.1 | AAK15698.1 | |||
CV809534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss | ABHD17A BTBD2 ONECUT3 ATP8B3 IZUMO4 MOB3A LOC100288123 MKNK2 AP3D1 CSNK1G2-AS1 ADAT3 SCAMP4 MIR1909 REXO1 CSNK1G2 KLF16 |
esv2669985 | Chr.19:1852046 - 1864993 on Build GRCh38 | Deletion | KLF16 |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | ABHD17A BTBD2 ONECUT3 ATP8B3 LOC100288123 APC2 PCSK4 RPS15 MUM1 CSNK1G2-AS1 UQCR11 DAZAP1 ADAT3 SCAMP4 REXO1 MBD3 MEX3D C19orf25 REEP6 GAMT TCF3 PLK5 NDUFS7 MIR1909 CSNK1G2 ADAMTSL5 KLF16 |
nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss | ABHD17A BTBD2 ATP8B3 IZUMO4 MOB3A LOC100288123 MKNK2 CSNK1G2-AS1 ADAT3 SCAMP4 MIR1909 REXO1 CSNK1G2 KLF16 |
nsv1160560 | Chr.19:1791100 - 1916393 on Build GRCh38 | Deletion | ABHD17A ATP8B3 ADAT3 SCAMP4 LOC100288123 MIR1909 REXO1 KLF16 |
nsv578338 | Chr.19:1847699 - 1869095 on Build GRCh38 | Loss | REXO1 KLF16 |
nsv578337 | Chr.19:1808864 - 1904613 on Build GRCh38 | Gain | ABHD17A ATP8B3 LOC100288123 MIR1909 REXO1 KLF16 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | ABHD17A BTBD2 ONECUT3 ATP8B3 LOC100288123 APC2 PCSK4 RPS15 MUM1 CSNK1G2-AS1 UQCR11 DAZAP1 ADAT3 SCAMP4 REXO1 MBD3 MEX3D C19orf25 REEP6 MKNK2 GAMT TCF3 PLK5 NDUFS7 MIR1909 CSNK1G2 ADAMTSL5 KLF16 |
esv2660894 | Chr.19:1854446 - 1872693 on Build GRCh38 | Deletion | KLF16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |