Assay Details
Target Gene Details
Entrez Gene ID: | 83855 |
Gene Name: | Kruppel like factor 16 |
Gene Aliases: |
BTEB4, DRRF, NSLP2 |
Location: |
Chr.19:1852399-1863565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF16 | NM_031918.3 | NP_114124.1 | ||
AF327440.1 | AAK15698.1 | |||
CV809534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss | REXO1 IZUMO4 BTBD2 MOB3A MKNK2 MIR1909 CSNK1G2-AS1 ATP8B3 ABHD17A CSNK1G2 KLF16 SCAMP4 AP3D1 ADAT3 LOC100288123 ONECUT3 |
esv2669985 | Chr.19:1852046 - 1864993 on Build GRCh38 | Deletion | KLF16 |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | PLK5 REXO1 MUM1 BTBD2 UQCR11 MIR1909 CSNK1G2-AS1 ABHD17A PCSK4 CSNK1G2 KLF16 ADAMTSL5 ADAT3 APC2 NDUFS7 GAMT MEX3D ATP8B3 DAZAP1 REEP6 SCAMP4 TCF3 MBD3 LOC100288123 ONECUT3 RPS15 C19orf25 |
nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss | REXO1 IZUMO4 BTBD2 MOB3A MKNK2 MIR1909 CSNK1G2-AS1 ATP8B3 ABHD17A CSNK1G2 KLF16 SCAMP4 ADAT3 LOC100288123 |
nsv1160560 | Chr.19:1791100 - 1916393 on Build GRCh38 | Deletion | ABHD17A KLF16 SCAMP4 REXO1 ADAT3 MIR1909 LOC100288123 ATP8B3 |
nsv578338 | Chr.19:1847699 - 1869095 on Build GRCh38 | Loss | KLF16 REXO1 |
nsv578337 | Chr.19:1808864 - 1904613 on Build GRCh38 | Gain | ABHD17A KLF16 REXO1 MIR1909 LOC100288123 ATP8B3 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | PLK5 REXO1 MUM1 BTBD2 UQCR11 MKNK2 MIR1909 CSNK1G2-AS1 ABHD17A PCSK4 CSNK1G2 KLF16 ADAMTSL5 ADAT3 APC2 NDUFS7 GAMT MEX3D ATP8B3 DAZAP1 REEP6 SCAMP4 TCF3 MBD3 LOC100288123 ONECUT3 RPS15 C19orf25 |
esv2660894 | Chr.19:1854446 - 1872693 on Build GRCh38 | Deletion | KLF16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |