Assay Details
Target Gene Details
Entrez Gene ID: | 4729 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit V2 |
Gene Aliases: |
CI-24k |
Location: |
Chr.18:9102623-9134345 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFV2 | NM_021074.4 | NP_066552.2 | ||
XM_017025782.1 | XP_016881271.1 | |||
AF147367.1 | ||||
AK291865.1 | ||||
AK312129.1 | ||||
AW051350.1 | ||||
BC001632.1 | AAH01632.1 | |||
BC017487.1 | AAH17487.1 | |||
BC071689.1 | AAH71689.1 | |||
BG742948.1 | ||||
CR456928.1 | CAG33209.1 | |||
M22538.1 | AAA75390.1 |
Target Gene Details
Entrez Gene ID: | 101927275 |
Gene Name: | NDUFV2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:9121265-9136720 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFV2-AS1 | NR_110771.1 | |||
NR_110772.1 | ||||
BI520375.1 | ||||
HY015092.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |