Assay Details
Target Gene Details
Entrez Gene ID: | 162394 |
Gene Name: | schlafen family member 5 |
Gene Aliases: |
- |
Location: |
Chr.17:35243026-35273650 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLFN5 | NM_144975.3 | NP_659412.3 | ||
XM_005257934.3 | XP_005257991.1 | |||
AK054668.1 | BAB70788.1 | |||
AK303299.1 | ||||
AK303632.1 | ||||
BC021238.1 | AAH21238.1 | |||
BP281007.1 | ||||
BX647942.1 | ||||
CD704489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422399 | Chr.17:35212221 - 35291582 on Build GRCh38 | Deletion | SLFN5 LOC105371932 |
esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss | SLFN5 FNDC8 SLC35G3 RAD51L3-RFFL NLE1 RAD51D UNC45B LOC105371932 |
esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss | LRRC37A8P HEATR9 AP2B1 GAS2L2 LOC105371933 PEX12 SLFN14 SNORD7 TAF15 CCL5 SLFN11 C17orf50 SLFN12L SLFN5 RDM1 MMP28 RASL10B SLFN12 SLFN13 LOC105371743 |
nsv574741 | Chr.17:35225256 - 35500115 on Build GRCh38 | Loss | SLFN5 LOC105371933 SLFN12 SLFN13 SLFN11 SLFN12L LOC105371932 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |