Assay Details
Target Gene Details
Entrez Gene ID: | 105371814 |
Gene Name: | uncharacterized LOC105371814 |
Gene Aliases: |
- |
Location: |
Chr.17:48874860-48908983 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371814 | NR_135674.1 | |||
CX873301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv960120 | Chr.17:48873588 - 48878124 on Build GRCh38 | Duplication | LOC105371814 |
nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss | SNF8 GIP ATP5G1 LOC105371814 UBE2Z CALCOCO2 |