Assay Details
Target Gene Details
Entrez Gene ID: | 23510 |
Gene Name: | potassium channel tetramerization domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.17:75032567-75065889 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD2 | NR_110835.1 | |||
AK307083.1 | ||||
AK311278.1 | ||||
DA408645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893044 | Chr.17:75032657 - 75048093 on Build GRCh38 | Loss | KCTD2 ATP5H |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |