Assay Details
Target Gene Details
Entrez Gene ID: | 114897 |
Gene Name: | C1q and tumor necrosis factor related protein 1 |
Gene Aliases: |
CTRP1, GIP, ZSIG37 |
Location: |
Chr.17:79022934-79049788 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1QTNF1 | NM_030968.3 | NP_112230.1 | ||
NM_153372.1 | NP_699203.1 | |||
NM_198594.2 | NP_940996.1 | |||
NR_049769.1 | ||||
XM_006721664.1 | XP_006721727.1 | |||
XM_006721666.1 | XP_006721729.1 | |||
AK055541.1 | BAB70947.1 | |||
AK172767.1 | BAD18750.1 | |||
AK292034.1 | ||||
AY358424.1 | AAQ88790.1 | |||
DA877694.1 | ||||
DA910618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470610 | Chr.17:78973800 - 79091723 on Build GRCh38 | Loss | RBFOX3 C1QTNF1-AS1 LGALS3BP CANT1 C1QTNF1 ENGASE |
nsv523650 | Chr.17:78933797 - 79165563 on Build GRCh38 | Loss | RBFOX3 C1QTNF1-AS1 LGALS3BP CANT1 C1QTNF1 ENGASE |
nsv1057193 | Chr.17:78921031 - 79481744 on Build GRCh38 | Gain | RBFOX3 C1QTNF1-AS1 LGALS3BP CANT1 C1QTNF1 ENGASE TIMP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |