Assay Details
Target Gene Details
Entrez Gene ID: | 100131439 |
Gene Name: | CD300 molecule like family member d |
Gene Aliases: |
CD300D, CLM-4, CMRF35-A4, CMRF35A4 |
Location: |
Chr.17:74579972-74592231 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD300LD | NM_001115152.1 | NP_001108624.1 | ||
AY358148.1 | AAQ88515.1 | |||
EF137868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761978 | Chr.17:73835799 - 74684915 on Build GRCh38 | Gain | GPR142 TTYH2 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB BTBD17 MGC16275 LOC100130520 DNAI2 CD300A GPRC5C KIF19 LOC101928343 RPL38 |
esv3641230 | Chr.17:73831787 - 74694861 on Build GRCh38 | Gain | GPR142 TTYH2 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB BTBD17 MGC16275 CD300LF LOC100130520 DNAI2 CD300A GPRC5C KIF19 LOC101928343 RPL38 |
dgv3282n100 | Chr.17:73814343 - 74719597 on Build GRCh38 | Gain | GPR142 TTYH2 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB BTBD17 MGC16275 CD300LF LOC100130520 DNAI2 CD300A GPRC5C KIF19 LOC101928343 LINC00469 RPL38 |
esv33995 | Chr.17:74261053 - 74732093 on Build GRCh38 | Loss | GPR142 TTYH2 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB BTBD17 CD300LF LOC100130520 DNAI2 CD300A GPRC5C KIF19 LOC101928343 |
nsv517569 | Chr.17:73837862 - 74685802 on Build GRCh38 | Gain+Loss | GPR142 TTYH2 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB BTBD17 MGC16275 LOC100130520 DNAI2 CD300A GPRC5C KIF19 LOC101928343 RPL38 |
dgv5768n54 | Chr.17:73826016 - 74706775 on Build GRCh38 | Gain | GPR142 TTYH2 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB BTBD17 MGC16275 CD300LF LOC100130520 DNAI2 CD300A GPRC5C KIF19 LOC101928343 LINC00469 RPL38 |
esv3573820 | Chr.17:73817932 - 74694378 on Build GRCh38 | Gain | GPR142 TTYH2 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB BTBD17 MGC16275 CD300LF LOC100130520 DNAI2 CD300A GPRC5C KIF19 LOC101928343 LINC00469 RPL38 |
esv3641244 | Chr.17:74527395 - 74686883 on Build GRCh38 | Gain | LOC100130520 CD300E C17orf77 CD300C RAB37 CD300LD CD300LB LOC101928343 |
dgv5774n54 | Chr.17:74428381 - 74673021 on Build GRCh38 | Gain | LOC100130520 CD300A CD300E C17orf77 CD300C GPRC5C RAB37 CD300LD CD300LB LOC101928343 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |