Assay Details
Target Gene Details
Entrez Gene ID: | 339230 |
Gene Name: | coiled-coil domain containing 137 |
Gene Aliases: |
RaRF |
Location: |
Chr.17:81666697-81673907 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC137 | NM_199287.2 | NP_954981.1 | ||
XM_011524738.2 | XP_011523040.1 | |||
XM_017024573.1 | XP_016880062.1 | |||
BC009369.2 | AAH09369.1 | |||
BM548065.1 | ||||
FJ649645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | TMEM105 LOC400627 FSCN2 CEP131 LOC101928855 LINC00482 SLC25A10 MIR6786 TSPAN10 ACTG1 AATK-AS1 LOC105371925 LOC105371921 LOC100130370 MIR338 C17orf89 MRPL12 NPLOC4 OXLD1 LOC100129503 BAHCC1 RPTOR BAIAP2 MIR3186 PDE6G LOC107985022 FAAP100 MIR1250 CCDC137 MIR3065 SLC38A10 CHMP6 LOC107985021 BAIAP2-AS1 ARL16 AATK TEPSIN HGS MIR657 MIR4740 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | TMEM105 RFNG ANAPC11 SLC25A10 CSNK1D FOXK2 AATK-AS1 WDR45B LOC105371925 TEX19 LOC105371944 B3GNTL1 PPP1R27 MRPL12 RAB40B FN3K TBCD LOC105376791 MIR3186 PDE6G FAAP100 MAFG NOTUM CCDC137 ZNF750 FASN SECTM1 TEPSIN PCYT2 GPS1 HGS STRA13 DUS1L PYCR1 P4HB MIR4740 CCDC57 FSCN2 CEP131 LINC00482 MIR6786 TSPAN10 ACTG1 LOC101929552 HEXDC LOC101929511 LOC100130370 LRRC45 C17orf89 DCXR ASPSCR1 SLC16A3 NPLOC4 ALYREF OGFOD3 OXLD1 UTS2R MIR4525 BAHCC1 FN3KRP ARHGDIA SIRT7 C17orf62 GCGR MIR6787 NPB CD7 SLC38A10 SNORD134 MCRIP1 LOC105376790 RAC3 MAFG-AS1 ARL16 AATK NARF MYADML2 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | FSCN2 RFNG ANAPC11 SLC25A10 MIR6786 TSPAN10 LRRC45 DCXR PPP1R27 ASPSCR1 MRPL12 NPLOC4 ALYREF OXLD1 ARHGDIA SIRT7 PDE6G GCGR FAAP100 NPB MAFG NOTUM CCDC137 MCRIP1 RAC3 MAFG-AS1 ARL16 PCYT2 GPS1 MYADML2 HGS STRA13 DUS1L PYCR1 P4HB |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | SLC25A10 MIR6786 TSPAN10 PDE6G GCGR CCDC137 MCRIP1 PPP1R27 ARL16 MRPL12 HGS P4HB OXLD1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |